Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107049790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145718.1 | XP_025001486.1 | 39 - 40 | 5646 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |