Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC770936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154711.1 | XP_025010479.1 | 4 - 5 | 3052 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |