Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il21r.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113509.1 | NP_001106981.1 | 1 - 2 | 198 | 123 | |
XM_021469048.1 | XP_021324723.1 | 1 - 2 | 198 | 123 | ||
XM_021469054.1 | XP_021324729.1 | 2 - 3 | 326 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() |