Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pon3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105122.2 | NP_001098592.2 | 1 - 2 | 119 | 105 | |
GenBank mRNA | BC152247.1 | - | 1 - 2 | 105 | ||
CB365732.1 | - | 1 - 2 | 105 | |||
EH548436.1 | - | 1 - 2 | 150 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |