Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cela1.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003737.1 | NP_001003737.1 | 1 - 2 | 48 | 84 | |
GenBank mRNA | BC078376.1 | - | 1 - 2 | 48 | 84 | |
BC165698.1 | - | 1 - 2 | 75 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |