Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tomm20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002698.1 | NP_001002698.1 | 2 - 3 | 273 | 59 | |
GenBank mRNA | BC076116.1 | - | 2 - 3 | 273 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |