Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079660.1 | NP_001073128.1 | 2 - 3 | 120 | 70 | |
XM_009305208.2 | XP_009303483.1 | 2 - 3 | 185 | 70 | ||
GenBank mRNA | BC127570.1 | - | 2 - 3 | 120 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |