Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181761.2 | NP_861426.1 | 11 - 12 | 882 | 60 | |
GenBank mRNA | AY178796.1 | - | 11 - 12 | 868 | 60 | |
BC056699.1 | - | 11 - 12 | 882 | 60 | ||
BC062531.1 | - | 11 - 12 | 874 | 60 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |