Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100765753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_035461208.1 | XP_035317099.1 | 9 - 10 | 1123 | 70 |
Set Membership: |
![]() ![]() |