Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495125.5 | XP_003495173.1 | 3 - 4 | 344 | 63 | |
GenBank mRNA | JI878139.1 | - | 3 - 4 | 354 | 63 | |
JP064052.1 | - | 3 - 4 | 354 | 63 |
Set Membership: |
![]() ![]() ![]() |