Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100764653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_035456022.1 | XP_035311913.1 | 8 - 9 | 1423 | 108 |
Set Membership: |
![]() ![]() ![]() |