Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc162p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_035438405.1 | XP_035294296.1 | 24 - 25 | 3524 | 96 |