Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022415321.1 | XP_022271029.1 | 3 | 1512 | 186 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |