Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16D2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063547.5 | NP_495948.1 | 1 | 306 | 70 |
Gene Symbol: jun-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383910.1 | 7 | 971 | 70 | ||
GenBank mRNA | EU553923.1 | - | 1 | 163 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |