Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02499020_g1
See other F49E2.5 GE Assays ›
Gene Symbol:
F49E2.5
Gene Name:
protein-coding
Gene Aliases:
CELE_F49E2.5
Chromosome Location:
Chr.X: 9543652 - 9560804 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02499020_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: F49E2.5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001029523.5
NP_001024694.1
13 - 14
2781
95
NM_001029524.6
NP_001024695.1
15 - 16
3003
95
NM_001373430.3
NP_001360758.1
17 - 18
3635
95
NM_001373432.4
NP_001360760.1
16 - 17
3077
95
NM_001373433.2
NP_001360761.1
14 - 15
2870
95
NM_001381054.2
NP_001368747.1
16 - 17
3497
95
NM_001381055.1
NP_001366703.1
11 - 12
2160
95
NM_001381056.1
NP_001366700.1
11 - 12
2583
95
NM_001381057.1
NP_001367097.1
10 - 11
2028
95
NM_001381058.1
NP_001368748.1
15 - 16
3192
95
NM_001381059.1
NP_001368749.1
14 - 15
3099
95
NM_001381060.1
NP_001368750.1
14 - 15
2637
95
NM_001381061.2
NP_001368751.1
13 - 14
2544
95
NM_001381062.1
NP_001368752.1
12 - 13
2427
95
NM_001381063.1
NP_001368753.1
11 - 12
2334
95
NM_001381064.1
NP_001366704.1
10 - 11
2390
95
NM_001381065.1
NP_001368754.1
10 - 11
1872
95
NM_001381066.1
NP_001368755.1
9 - 10
1835
95
NM_001381067.1
NP_001367224.1
8 - 9
1590
95
NM_001383624.1
13 - 14
3188
95
NM_001383625.1
13 - 14
4018
95
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Detect Genomic DNA
Caenorhabditis elegans
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0