Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047695.5 | NP_001041160.1 | 9 - 10 | 1989 | 81 | |
NM_001047696.3 | NP_001041161.1 | 8 - 9 | 1953 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() |