Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069208.3 | NP_501609.1 | 1 - 2 | 126 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |