Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T28D6.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067060.7 | NP_499461.1 | 4 - 5 | 1787 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |