Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066859.5 | NP_499260.1 | 2 - 3 | 296 | 136 |
Set Membership: |
![]() ![]() ![]() ![]() |