Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065836.10 | NP_498237.2 | 1 - 2 | 62 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() |