Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ce02444346_m1
          See other DAF-2 GE Assays ›
          Gene Symbol:
          daf-2
          Gene Name:
          protein-coding
          Gene Aliases:
          CELE_Y55D5A.5
          Chromosome Location:
          Chr.III: 2994514 - 3040860 on Build WBcel235
          Species:
          Caenorhabditis elegans
          Species Specific ID (Flybase ID):
          -
          Assay ID Ce02444346_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: daf-2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001312987.3 NP_001299916.1 5 583 142
          NM_001312988.3 NP_001299917.1 3 - 4 265 142
          NM_001312989.3 NP_001299918.1 2 - 3 178 142
          NM_065249.6 NP_497650.4 5 - 6 583 142
          NR_132448.1 4 - 5 406 142

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          reproduction
          immune system process
          carbohydrate metabolic process
          protein phosphorylation
          protein import into nucleus
          response to oxidative stress
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          insulin receptor signaling pathway
          determination of adult lifespan
          response to UV
          heat acclimation
          regulation of gene expression
          positive regulation of gene expression
          negative regulation of gene expression
          regulation of lipid storage
          negative regulation of lipid storage
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          peptidyl-tyrosine phosphorylation
          larval feeding behavior
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of kinase activity
          positive regulation of multicellular organism growth
          dauer larval development
          regulation of development, heterochronic
          glucose homeostasis
          dauer exit
          positive regulation of MAPK cascade
          innate immune response
          positive regulation of synaptic assembly at neuromuscular junction
          positive regulation of transcription by RNA polymerase II
          protein autophosphorylation
          positive regulation of protein kinase B signaling
          regulation of dauer larval development
          positive regulation of neuromuscular synaptic transmission
          negative regulation of protein localization to nucleus
          regulation of response to reactive oxygen species
          cellular response to salt
          regulation of eating behavior
          regulation of dauer entry
          negative regulation of dauer entry
          regulation of autophagosome assembly
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          insulin-activated receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          peptide hormone binding
          protein kinase binding
          SH2 domain binding
          insulin receptor substrate binding
          metal ion binding
          PTB domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0