Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060438.7 | NP_492839.4 | 16 - 17 | 6313 | 145 | |
GenBank mRNA | AB297384.1 | - | 16 - 17 | 6299 | 145 |
Set Membership: |
![]() ![]() ![]() ![]() |