Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060438.7 | NP_492839.4 | 14 - 15 | 4700 | 100 | |
GenBank mRNA | AB297384.1 | - | 14 - 15 | 4686 | 100 |
Set Membership: |
![]() ![]() ![]() |