Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060438.7 | NP_492839.4 | 10 - 11 | 2002 | 79 | |
GenBank mRNA | AB297384.1 | - | 10 - 11 | 1988 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() |