Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192635.1 | NP_001179564.1 | 1 - 2 | 649 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |