Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JSP.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040498.1 | NP_001035588.1 | 2 - 3 | 334 | 157 | |
GenBank mRNA | BC105263.1 | - | 2 - 3 | 334 | 157 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |