Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083411.2 | NP_001076880.2 | 10 - 11 | 1209 | 137 | |
GenBank mRNA | BC123768.1 | - | 9 - 10 | 1657 | 137 | |
BT030692.1 | - | 10 - 11 | 1209 | 137 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |