Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025319.2 | NP_001020490.1 | 3 - 4 | 467 | 86 | |
GenBank mRNA | BC149346.1 | - | 3 - 4 | 577 | 86 | |
DQ078267.1 | - | 3 - 4 | 572 | 86 | ||
DQ085625.1 | - | 3 - 4 | 467 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() |