Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Bt03224502_m1
          See other RHOA GE Assays ›
          Gene Symbol:
          RHOA
          Gene Name:
          ras homolog family member A
          Gene Aliases:
          ARHA
          Chromosome Location:
          Chr.22: 50701226 - 50751133 on Build ARS-UCD1.2
          Species:
          Bovine
          Species Specific ID (Flybase ID):
          -
          Assay ID Bt03224502_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RHOA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176645.3 NP_788818.1 5 - 6 643 78
          GenBank mRNA BC102880.1 - 5 - 6 643 78
          M27278.1 - 5 - 6 607 78

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          angiotensin-mediated vasoconstriction involved in regulation of systemic arterial blood pressure
          alpha-beta T cell lineage commitment
          regulation of systemic arterial blood pressure by endothelin
          regulation of transcription by RNA polymerase II
          cell-matrix adhesion
          Rho protein signal transduction
          skeletal muscle tissue development
          negative regulation of cell-substrate adhesion
          regulation of neuron projection development
          cell migration
          cerebral cortex cell migration
          forebrain radial glial cell differentiation
          regulation of cell migration
          androgen receptor signaling pathway
          cytoplasmic microtubule organization
          actin cytoskeleton reorganization
          positive regulation of cytokinesis
          negative regulation of intracellular steroid hormone receptor signaling pathway
          regulation of osteoblast proliferation
          cell junction assembly
          substrate adhesion-dependent cell spreading
          Roundabout signaling pathway
          cleavage furrow formation
          apolipoprotein A-I-mediated signaling pathway
          odontogenesis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          stress fiber assembly
          apical junction assembly
          beta selection
          negative regulation of neuron apoptotic process
          endothelial cell migration
          ossification involved in bone maturation
          wound healing, spreading of cells
          establishment of epithelial cell apical/basal polarity
          positive regulation of neuron differentiation
          negative regulation of cell size
          positive regulation of alpha-beta T cell differentiation
          negative chemotaxis
          positive regulation of stress fiber assembly
          positive regulation of lipase activity
          trabecula morphogenesis
          regulation of microtubule cytoskeleton organization
          cellular response to lipopolysaccharide
          positive regulation of podosome assembly
          positive regulation of protein serine/threonine kinase activity
          negative regulation of cell migration involved in sprouting angiogenesis
          mitotic spindle assembly
          negative regulation of oxidative phosphorylation
          endothelial tube lumen extension
          positive regulation of NIK/NF-kappaB signaling
          skeletal muscle satellite cell migration
          negative regulation of reactive oxygen species biosynthetic process
          mitotic cleavage furrow formation
          positive regulation of vascular associated smooth muscle contraction
          positive regulation of leukocyte adhesion to vascular endothelial cell
          regulation of modification of postsynaptic actin cytoskeleton
          cellular response to chemokine
          regulation of neural precursor cell proliferation
          positive regulation of T cell migration
          GTPase activity
          GTP binding
          myosin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0