Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G09660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117034.1 | NP_192704.1 | 2 - 3 | 952 | 163 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |