Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2A-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129811.4 | NP_565974.1 | 10 - 11 | 1062 | 157 | |
NM_201943.2 | NP_973672.1 | 8 - 9 | 921 | 157 | ||
GenBank mRNA | M96841.1 | - | 10 - 11 | 1059 | 157 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |