Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124144.1 | NP_001117616.1 | 4 - 5 | 293 | 76 | |
NM_001334806.1 | NP_001322357.1 | 5 - 6 | 337 | 76 | ||
NM_001334807.1 | NP_001322358.1 | 4 - 5 | 417 | 76 | ||
NM_106444.3 | NP_177918.2 | 4 - 5 | 417 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |