Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EC1.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125868.4 | NP_201277.3 | 1 | 441 | 90 | |
GenBank mRNA | BT029327.1 | - | 1 | 421 | 90 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |