Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP78A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121034.2 | NP_196559.1 | 1 - 2 | 1294 | 78 | |
GenBank mRNA | AK227001.1 | - | 1 - 2 | 1298 | 78 | |
AY136401.1 | - | 1 - 2 | 1294 | 78 | ||
BT000225.1 | - | 1 - 2 | 998 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |