Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338439.1 | NP_001326561.1 | 8 - 9 | 1114 | 107 | |
NM_112890.5 | NP_188634.1 | 8 - 9 | 922 | 107 | ||
GenBank mRNA | AY050925.1 | - | 8 - 9 | 764 | 107 | |
AY117216.1 | - | 8 - 9 | 699 | 107 |
Set Membership: |
![]() ![]() ![]() ![]() |