Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR3;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112469.3 | NP_188220.1 | 7 - 8 | 1141 | 137 | |
GenBank mRNA | AB054645.1 | - | 7 - 8 | 1141 | 137 |
Set Membership: |
![]() ![]() ![]() ![]() |