Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177360.1 | NP_001170831.1 | 1 | 695 | ||
NM_207220.2 | NP_997103.1 | 1 | 695 | |||
GenBank mRNA | AK039612.1 | - | 1 | 195 | ||
AK142928.1 | - | 1 | 293 | |||
BC048078.1 | - | 2 | 567 | 6308873 | ||
CX200961.1 | - | 1 | 695 |