Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163754.1 | NP_001157226.1 | 32 | 3665 | ||
GenBank mRNA | AK031191.1 | - | 22 | 2574 | ||
AK048759.1 | - | 8 | 2006 | |||
AK122385.1 | - | 17 | 4353 | |||
AK149265.1 | - | 9 | 836 | |||
BC035944.1 | - | 23 | 2655 | |||
BC052505.1 | - | 7 | 763 | |||
BC057872.1 | - | 23 | 2683 | 5294131 |