Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002841.1 | NP_001002841.1 | 4, 5 | 450 | ||
NM_002476.2 | NP_002467.1 | 3, 4 | 397 | |||
XM_005257391.4 | XP_005257448.1 | 4, 5 | 377 | |||
XM_011524839.2 | XP_011523141.2 | 4, 5 | 537 | |||
XM_017024683.1 | XP_016880172.1 | 4, 5 | 524 | |||
XM_017024684.1 | XP_016880173.1 | 3, 4 | 541 | |||
GenBank mRNA | AF116676.2 | - | 749 | |||
BC030228.1 | - | 3, 4 | 386 | 5226518 | ||
BU658678.1 | - | 4, 5 | 456 | |||
M20641.1 | - | 1 | 13 | |||
M24121.1 | - | 3, 4 | 384 | |||
M36172.1 | - | 3, 4 | 378 | |||
X13955.1 | - | 3, 4 | 378 |