Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027901.2 | NP_082177.2 | 13 | 2029 | ||
GenBank mRNA | AK004895.1 | - | 13 | 1807 | ||
AK012095.1 | - | 6 | 626 | |||
AK043944.1 | - | 13 | 1796 | |||
AK083585.1 | - | 13 | 2031 | |||
AK083636.1 | - | 12 | 1784 | |||
AK129033.1 | - | 12 | 1779 | |||
AK138760.1 | - | 1798 | ||||
AK149771.1 | - | 14 | 1926 | |||
AK151300.1 | - | 13 | 1794 | |||
AK152439.1 | - | 13 | 1794 | |||
AK159938.1 | - | 13 | 1797 | |||
AK193271.1 | - | 4 | 233 | |||
AK195721.1 | - | 4 | 218 | |||
BC026573.1 | - | 5 | 347 | |||
BC034369.1 | - | 13 | 1807 | 3994677 | ||
BC043100.1 | - | 13 | 2017 |