Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177437.1 | NP_001170908.1 | 16 | 1981 | ||
GenBank mRNA | AK006048.1 | - | 16 | 1926 | ||
BC111897.1 | - | 13 | 1537 | 40061435 |