Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTAGE5 SIRNA Assays ›
          Gene Symbol:
          CTAGE5
          Gene Name:
          CTAGE family member 5, ER export factor
          Gene Aliases:
          MEA6, MGEA, MGEA11, MGEA6
          Chromosome Location:
          Chr.14: 39265264 - 39351193 on Build GRCh38
          UniGene ID:
          Hs.509200
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8744

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTAGE5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001247988.1 NP_001234917.1 2 267
          NM_001247989.1 NP_001234918.1 2 436
          NM_005930.3 NP_005921.2 2 436
          NM_203354.2 NP_976229.1 2 187
          NM_203355.2 NP_976230.1 2 436
          NM_203356.2 NP_976231.1 2 267
          XM_005267648.4 XP_005267705.1 2 147
          XM_006720148.2 XP_006720211.1 2 149
          XM_011536775.2 XP_011535077.1 2 487
          XM_011536778.2 XP_011535080.1 2 175
          XM_017021314.1 XP_016876803.1 2 488
          XM_017021315.1 XP_016876804.1 2 189
          XM_017021316.1 XP_016876805.1 2 176
          XM_017021317.1 XP_016876806.1 2 597
          XM_017021318.1 XP_016876807.1 2 218
          XM_017021319.1 XP_016876808.1 2 154
          XM_017021320.1 XP_016876809.1 2 222
          XM_017021321.1 XP_016876810.1 2 488
          XM_017021322.1 XP_016876811.1 2 195
          XM_017021325.1 XP_016876814.1 2 155
          XM_017021330.1 XP_016876819.1 2 97
          GenBank mRNA AB209528.1 - 1 745
          AF338233.1 - 2 281
          AF338234.1 - 2 314
          AK055791.1 - 2 556
          AK091252.1 - 2 645
          AK298935.1 - 2 168
          AK315254.1 - 2 157
          AL832233.1 - 2 150
          AL833415.1 - 2 96
          BC029513.1 - 2 154
          BC039017.1 - 2 193
          BC064355.1 - 2 382 4803802
          BX367676.2 - 2 386
          BX640994.1 - 2 168
          BX648014.1 - 2 168
          CD512382.1 - 2 174
          CV809109.1 - 2 458
          DA072098.1 - 2 187
          U73682.1 - 2 146
          U94780.1 - 2 414

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ER to Golgi vesicle-mediated transport
          protein exit from endoplasmic reticulum
          cargo loading into vesicle
          positive regulation of catalytic activity
          protein localization to endoplasmic reticulum exit site
          regulation of guanyl-nucleotide exchange factor activity
          protein binding
          enzyme activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-956b84784-lf4xt:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0