Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006166.1 | - | 3 | 487 | ||
AK009929.1 | - | 3 | 323 | |||
AK011393.1 | - | 3 | 481 | |||
AK011848.1 | - | 3 | 487 | |||
AK017100.1 | - | 3 | 349 | |||
AK017322.1 | - | 3 | 481 | |||
AK029148.1 | - | 483 | ||||
AK030275.1 | - | 481 | ||||
AK147270.1 | - | 494 | ||||
AK170765.1 | - | 3 | 487 | |||
BC026913.1 | - | 3 | 313 |