Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020561.2 | NP_065586.3 | 2 | 286 | ||
GenBank mRNA | AK002455.1 | - | 2 | 225 | ||
AK151683.1 | - | 2 | 172 | |||
AK152282.1 | - | 2 | 187 | |||
AK185060.1 | - | 1 | 32 | |||
AK194580.1 | - | 1 | 32 | |||
AK198108.1 | - | 202 | ||||
AK205531.1 | - | 1 | 32 | |||
BC139288.1 | - | 2 | 178 | 8862310 | ||
BC139289.1 | - | 2 | 178 | 8862311 | ||
Y08135.1 | - | 2 | 240 |