Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019822.3 | NP_062796.2 | 2 | 229 | ||
GenBank mRNA | AF225959.1 | - | 2 | 145 | ||
AK010919.1 | - | 2 | 206 | |||
AK082084.1 | - | 2 | 212 | |||
AK143843.1 | - | 2 | 204 | |||
AK145474.1 | - | 2 | 191 | |||
AK145792.1 | - | 2 | 220 | |||
AK150091.1 | - | 2 | 182 | |||
AK166878.1 | - | 2 | 218 | |||
AK207110.1 | - | 1 | 49 | |||
BC008974.1 | - | 2 | 229 | 3586698 | ||
BC019746.1 | - | 2 | 158 | 4188750 | ||
BC031517.1 | - | 2 | 172 | 4008987 |