Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009459.3 | NP_033485.1 | 3, 4 | 639 | ||
GenBank mRNA | AK131688.1 | - | 3, 4 | 617 | ||
BC008517.1 | - | 3, 4 | 445 | 3600233 | ||
U19854.1 | - | 3, 4 | 368 |