Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042674.2 | NP_001036139.1 | 3, 4 | 516 | ||
NM_001042675.2 | NP_001036140.1 | 3, 4 | 516 | |||
NM_001286168.1 | NP_001273097.1 | 3, 4 | 516 | |||
NM_019733.3 | NP_062707.1 | 3, 4 | 516 | |||
GenBank mRNA | AF148511.1 | - | 3, 4 | 465 | ||
AK012265.1 | - | 3, 4 | 459 | |||
AK012586.1 | - | 3, 4 | 459 | |||
AK041118.1 | - | 3, 4 | 460 | |||
AK079238.1 | - | 3, 4 | 459 | |||
BB637557.1 | - | 3, 4 | 460 | |||
BC030397.1 | - | 3, 4 | 477 | 4225078 | ||
CK030989.1 | - | 3, 4 | 494 |