Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008791.2 | NP_032817.1 | 3 | 324 | ||
GenBank mRNA | BC061164.1 | - | 3 | 324 | 6593949 | |
X17320.1 | - | 3 | 163 |