Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008681.2 | NP_032707.2 | 10, 11 | 865 | ||
GenBank mRNA | AA795988.1 | - | 2, 3 | 158 | ||
AK004548.1 | - | 10, 11 | 865 | |||
AK098075.1 | - | 10, 11 | 864 | |||
AK161860.1 | - | 10, 11 | 867 | |||
AK165007.1 | - | 10, 11 | 865 | |||
AK170281.1 | - | 10, 11 | 866 | |||
AK170439.1 | - | 10, 11 | 864 | |||
AK170636.1 | - | 10, 11 | 864 | |||
AK170809.1 | - | 10, 11 | 865 | |||
BC015282.1 | - | 10, 11 | 820 | 4237943 | ||
BC071235.1 | - | 10, 11 | 873 | 4242075 | ||
BC141246.1 | - | 10, 11 | 756 | 9055858 | ||
BC171968.1 | - | 10, 11 | 756 | 9054254 | ||
CA320478.1 | - | 7, 8 | 608 | |||
U52073.1 | - | 10, 11 | 838 | |||
U60593.1 | - | 10, 11 | 863 |