Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008209.4 | NP_032235.1 | 3 | 493 | ||
GenBank mRNA | AF010448.1 | - | 3 | 404 | ||
AF010449.1 | - | 3 | 219 | |||
AF010450.1 | - | 3 | 219 | |||
AF010452.1 | - | 3 | 219 | |||
AF010453.1 | - | 219 | ||||
AK041171.1 | - | 2 | 1190 | |||
AK042119.1 | - | 3 | 334 | |||
AK137171.1 | - | 3 | 334 | |||
AK144040.1 | - | 3 | 438 | |||
AK162712.1 | - | 3 | 451 | |||
BC026137.1 | - | 3 | 521 | 4017492 | ||
U94989.1 | - | 3 | 433 |